Product Details
- SNP ID
-
rs10436957
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
45
- Location
-
Chr.1:15441808 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCACCTTAGCCTCCCGAGTAGCTG[A/G]GTCTATAAGCACACGCCACCACGCA
- Phenotype
-
MIM: 601405
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTRC
PubMed Links
Gene Details
- Gene
- CTRC
- Gene Name
- chymotrypsin C
View Full Product Details