Product Details

SNP ID
rs11622650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24407052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGCTCTGTTGCCTCTGGGCA[A/G]CAGAGTTACTATGATTAGGGGTACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NYNRIN PubMed Links

Gene Details

Gene
NYNRIN
Gene Name
NYN domain and retroviral integrase containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025081.2 Intron NP_079357.2
XM_011537016.2 Intron XP_011535318.1

View Full Product Details