Product Details
- SNP ID
-
rs35715
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:100576629 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTTTTAATTAGAGTTTAGTTGTCA[C/T]AGAGAAATGCTTCAGAAGTAATGTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GAS2L3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs113215628] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GAS2L3
- Gene Name
- growth arrest specific 2 like 3
- Gene
- PIGAP1
- Gene Name
- phosphatidylinositol glycan anchor biosynthesis class A pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details