Product Details
- SNP ID
-
rs852933
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:71408659 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCCTGAAGGCTATGATTCTGTCTC[A/G]ATCACTGCTGTACCTACTGGAGCAC
- Phenotype
-
MIM: 612329
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC00472
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76297264] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC00472
- Gene Name
- long intergenic non-protein coding RNA 472
There are no transcripts associated with this gene.
- Gene
- MIR30A
- Gene Name
- microRNA 30a
There are no transcripts associated with this gene.
View Full Product Details