Product Details

SNP ID
rs34648990
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:52084522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGGCTGGGCAACAGCACTGGTCA[A/G]AAGTTCCGACAGCTGACCACCCTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP64 PubMed Links

Gene Details

Gene
ZFP64
Gene Name
ZFP64 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319146.1 1964 UTR 3 NP_001306075.1
NM_018197.2 1964 Intron NP_060667.2
NM_022088.4 1964 Intron NP_071371.3
NM_199426.1 1964 Intron NP_955458.1
NM_199427.2 1964 UTR 3 NP_955459.2
XM_005260449.1 1964 Intron XP_005260506.1
XM_006723822.1 1964 Intron XP_006723885.1
XM_011528900.1 1964 Intron XP_011527202.1
XM_017027945.1 1964 UTR 3 XP_016883434.1
XM_017027946.1 1964 Intron XP_016883435.1
XM_017027947.1 1964 UTR 3 XP_016883436.1
XM_017027948.1 1964 Intron XP_016883437.1
XM_017027949.1 1964 Intron XP_016883438.1

View Full Product Details