Product Details

SNP ID
rs3784828
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:1529150 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTTGTGCCTGGTTTTACATGTACT[C/T]GGGTGAATTCCACGGACGCATTCCA
Phenotype
MIM: 614620 MIM: 611002
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IFT140 PubMed Links

Gene Details

Gene
IFT140
Gene Name
intraflagellar transport 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014714.3 994 Intron NP_055529.2
XM_005255725.4 994 Intron XP_005255782.1
XM_005255726.3 994 Intron XP_005255783.1
XM_006720990.3 994 Intron XP_006721053.1
XM_006720991.3 994 Intron XP_006721054.1
XM_006720992.3 994 UTR 5 XP_006721055.1
XM_011522766.2 994 Intron XP_011521068.1
XM_011522767.2 994 Intron XP_011521069.1
XM_011522769.2 994 Intron XP_011521071.1
XM_011522771.2 994 Intron XP_011521073.1
XM_011522772.2 994 Intron XP_011521074.1
XM_017023910.1 994 Intron XP_016879399.1
XM_017023911.1 994 Intron XP_016879400.1
Gene
LOC105371047
Gene Name
uncharacterized LOC105371047
There are no transcripts associated with this gene.

Gene
TMEM204
Gene Name
transmembrane protein 204
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256541.1 994 Intron NP_001243470.1
NM_024600.5 994 Intron NP_078876.2

View Full Product Details