Product Details

SNP ID
rs11919970
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:185192345 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TCCAGTTGGGGAATATCAGGGTTCT[G/T]CCTGTAATATTTCTGCAATTTCTCT
Phenotype
MIM: 607037
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EHHADH PubMed Links

Gene Details

Gene
EHHADH
Gene Name
enoyl-CoA, hydratase/3-hydroxyacyl CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166415.1 2111 Missense Mutation AAG,CAG K589Q NP_001159887.1
NM_001966.3 2111 Missense Mutation AAG,CAG K685Q NP_001957.2
XM_006713525.1 2111 Missense Mutation AAG,CAG K477Q XP_006713588.1
XM_011512517.1 2111 Missense Mutation AAG,CAG K589Q XP_011510819.1
XM_017005839.1 2111 Missense Mutation AAG,CAG K477Q XP_016861328.1
Gene
EHHADH-AS1
Gene Name
EHHADH antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details