Product Details

SNP ID
rs4819201
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.21:46106805 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCAAAACCAAAACTGTAATTTTATC[A/G]TTTCATTTGATAGCACAAGCTTTCT
Phenotype
MIM: 120240
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL6A2 PubMed Links

Gene Details

Gene
COL6A2
Gene Name
collagen type VI alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001849.3 Intron NP_001840.3
NM_058174.2 Intron NP_478054.2
NM_058175.2 Intron NP_478055.2
XM_011529451.1 Intron XP_011527753.1

View Full Product Details