Product Details

SNP ID
rs7532195
Assay Type
Functionally Tested
NCBI dbSNP Submissions
44
Location
Chr.1:93151846 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAGCATTTACAAATGACTGGCAG[G/T]TAATTTTCTTTGATAATTTTTATTA
Phenotype
MIM: 616876
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
TMED5 PubMed Links

Gene Details

Gene
TMED5
Gene Name
transmembrane p24 trafficking protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167830.1 4010 UTR 3 NP_001161302.1
NM_016040.4 4010 UTR 3 NP_057124.3

View Full Product Details