Product Details

SNP ID
rs2391323
Assay Type
Functionally tested
NCBI dbSNP Submissions
41
Location
Chr.1:93891848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAATGTTAGCTATTTTCATGTGC[C/G]AGGTACTTTTCTGAGTACTTTACAT
Phenotype
MIM: 601176
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GCLM PubMed Links

Gene Details

Gene
GCLM
Gene Name
glutamate-cysteine ligase modifier subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308253.1 Intron NP_001295182.1
NM_002061.3 Intron NP_002052.1
XM_011541261.2 Intron XP_011539563.1
XM_017001056.1 Intron XP_016856545.1
XM_017001057.1 Intron XP_016856546.1

View Full Product Details