Product Details
- SNP ID
-
rs36098342
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:78103566 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGGATGCCTCTGATCCATCCTAA[C/G]CCGACATTCCCTTGATCTCTCCTGA
- Phenotype
-
MIM: 605131
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
WWOX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74884323] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WWOX
- Gene Name
- WW domain containing oxidoreductase
View Full Product Details