Product Details

SNP ID
rs7197900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:78102685 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCGACCTTGTACTAGTCACCTTG[C/T]TTCTCTGCTTCTTCGTGCTTAGCCT
Phenotype
MIM: 605131
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WWOX PubMed Links

Gene Details

Gene
WWOX
Gene Name
WW domain containing oxidoreductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291997.1 Intron NP_001278926.1
NM_016373.3 Intron NP_057457.1
NM_130791.3 Intron NP_570607.1
XM_011523101.2 Intron XP_011521403.1
XM_011523103.2 Intron XP_011521405.1
XM_011523104.2 Intron XP_011521406.1
XM_011523105.2 Intron XP_011521407.1
XM_017023278.1 Intron XP_016878767.1
XM_017023279.1 Intron XP_016878768.1

View Full Product Details