Product Details
- SNP ID
-
rs2867794
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:44114513 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATGAATGAGTGAATGAAGCCCTCA[A/G]TGCTCACCTGACAGAGAGCAGGTGC
- Phenotype
-
MIM: 605267
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
JPH2
PubMed Links
Gene Details
- Gene
- JPH2
- Gene Name
- junctophilin 2
View Full Product Details