Product Details
- SNP ID
-
rs12008084
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:136675035 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGCCATGGCCTTGTTGAAAATTTGC[A/G]CTGGTGCAGTAGGCTTCGATCACTT
- Phenotype
-
MIM: 300267
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF6
PubMed Links
Gene Details
- Gene
- ARHGEF6
- Gene Name
- Rac/Cdc42 guanine nucleotide exchange factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306177.1 |
2874 |
Silent Mutation |
AGC,AGT |
S515S |
NP_001293106.1 |
NM_004840.2 |
2874 |
Silent Mutation |
AGC,AGT |
S669S |
NP_004831.1 |
XM_005262499.2 |
2874 |
Silent Mutation |
AGC,AGT |
S620S |
XP_005262556.1 |
XM_011531412.2 |
2874 |
Silent Mutation |
AGC,AGT |
S696S |
XP_011529714.1 |
XM_011531413.1 |
2874 |
Silent Mutation |
AGC,AGT |
S673S |
XP_011529715.1 |
XM_011531414.1 |
2874 |
Silent Mutation |
AGC,AGT |
S666S |
XP_011529716.1 |
XM_011531415.2 |
2874 |
Silent Mutation |
AGC,AGT |
S639S |
XP_011529717.1 |
XM_011531416.2 |
2874 |
Silent Mutation |
AGC,AGT |
S542S |
XP_011529718.1 |
XM_011531417.1 |
2874 |
Silent Mutation |
AGC,AGT |
S519S |
XP_011529719.1 |
XM_017029975.1 |
2874 |
Silent Mutation |
AGC,AGT |
S700S |
XP_016885464.1 |
XM_017029976.1 |
2874 |
Silent Mutation |
AGC,AGT |
S466S |
XP_016885465.1 |
View Full Product Details