Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001195263.1 | Intron | NP_001182192.1 | |||
NM_024895.4 | Intron | NP_079171.1 | |||
XM_005270165.2 | Intron | XP_005270222.1 | |||
XM_011540177.2 | Intron | XP_011538479.1 | |||
XM_011540178.2 | Intron | XP_011538480.1 | |||
XM_011540179.2 | Intron | XP_011538481.1 | |||
XM_011540181.2 | Intron | XP_011538483.1 | |||
XM_011540182.2 | Intron | XP_011538484.1 | |||
XM_011540183.2 | Intron | XP_011538485.1 | |||
XM_017016667.1 | Intron | XP_016872156.1 | |||
XM_017016668.1 | Intron | XP_016872157.1 |