Product Details

SNP ID
rs6675915
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:201041734 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCCGTGACTCTAGGGCTGACGTT[C/T]CCACCACTGTGCCCCAGCCAGCCCT
Phenotype
MIM: 114208
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CACNA1S PubMed Links

Gene Details

Gene
CACNA1S
Gene Name
calcium voltage-gated channel subunit alpha1 S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000069.2 Intron NP_000060.2
XM_005245478.3 Intron XP_005245535.1
Gene
LOC101929305
Gene Name
uncharacterized LOC101929305
There are no transcripts associated with this gene.

View Full Product Details