Product Details

SNP ID
rs34015581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71474326 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAATAGCCTGGCATTTCTCAGTT[A/T]AAGGCCTAGAGTAGCATCCATTCCA
Phenotype
MIM: 194525
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF19 PubMed Links

Gene Details

Gene
ZNF19
Gene Name
zinc finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006961.3 2490 UTR 3 NP_008892.2

View Full Product Details