Product Details

SNP ID
rs12461099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41861895 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCAATCTTCCAATAATAATTTA[C/T]GTACAGTCCCTGACTAGTGTTAGTT
Phenotype
MIM: 614806 MIM: 603474
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DMRTC2 PubMed Links

Gene Details

Gene
DMRTC2
Gene Name
DMRT like family C2
There are no transcripts associated with this gene.

Gene
MIR6797
Gene Name
microRNA 6797
There are no transcripts associated with this gene.

Gene
RPS19
Gene Name
ribosomal protein S19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001022.3 Intron NP_001013.1
NM_001321483.1 Intron NP_001308412.1
NM_001321484.1 Intron NP_001308413.1
NM_001321485.1 Intron NP_001308414.1
XM_017027113.1 Intron XP_016882602.1

View Full Product Details