Product Details
- SNP ID
-
rs10186535
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:206653460 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GATGTTAGATGGAACACTTGTGAAA[C/T]TGGCAGTCCAGAGAAAATCACAATG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DYTN
PubMed Links
Gene Details
- Gene
- DYTN
- Gene Name
- dystrotelin
- Gene
- LOC200726
- Gene Name
- hCG1657980
There are no transcripts associated with this gene.
View Full Product Details