Product Details

SNP ID
rs10042075
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:55846898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGCTCAAGCAGGTGCCCTATTAT[A/G]CTTTCTGGAATTTAAAAATAAACTT
Phenotype
MIM: 609510
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL31RA PubMed Links

Gene Details

Gene
IL31RA
Gene Name
interleukin 31 receptor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242636.1 Intron NP_001229565.1
NM_001242637.1 Intron NP_001229566.1
NM_001242638.1 Intron NP_001229567.1
NM_001242639.1 Intron NP_001229568.1
NM_001297570.1 Intron NP_001284499.1
NM_001297572.1 Intron NP_001284501.1
NM_139017.5 Intron NP_620586.3
XM_011543142.2 Intron XP_011541444.1
XM_011543143.2 Intron XP_011541445.1
XM_011543144.2 Intron XP_011541446.1
XM_011543145.2 Intron XP_011541447.1

View Full Product Details