Product Details

SNP ID
rs13162617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:52791236 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGTTTCATAAAGTGACAGTTTAG[A/G]GAGGCCTTAAAGGAACAAGAAAAGA
Phenotype
MIM: 192968 MIM: 605757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ITGA1 PubMed Links

Gene Details

Gene
ITGA1
Gene Name
integrin subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181501.1 Intron NP_852478.1
Gene
PELO
Gene Name
pelota homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015946.4 Intron NP_057030.3

View Full Product Details