Product Details

SNP ID
hCV2844089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69582573 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCCTAACAGCTTCATTGTTTTGC[A/G]TGTGCATATCTAGTTGCCTGAGCAC
Phenotype
MIM: 604708
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFAT5 PubMed Links

Gene Details

Gene
NFAT5
Gene Name
nuclear factor of activated T-cells 5, tonicity-responsive
There are no transcripts associated with this gene.

View Full Product Details