Product Details

SNP ID
rs1058747
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:71645301 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGCTTTTTCTCAATCTAGGAAGTG[C/T]TTACCCCAACTATGGGGCAAAAGTC
Phenotype
MIM: 614094 MIM: 611066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MARVELD3 PubMed Links

Gene Details

Gene
MARVELD3
Gene Name
MARVEL domain containing 3
There are no transcripts associated with this gene.

Gene
PHLPP2
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289003.1 7940 UTR 3 NP_001275932.1
NM_015020.3 7940 UTR 3 NP_055835.2

View Full Product Details