Product Details
- SNP ID
-
rs11640135
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:71847127 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTGCACCTTGCCAATCTTAAGAG[A/G]GAAATGTTGCATTCGGTTTGGGTTG
- Phenotype
-
MIM: 614301
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATXN1L
PubMed Links
Gene Details
- Gene
- ATXN1L
- Gene Name
- ataxin 1 like
View Full Product Details