Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_024516.3 | 154 | UTR 5 | NP_078792.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001256442.1 | 154 | Intron | NP_001243371.1 | ||
NM_001256443.1 | 154 | Intron | NP_001243372.1 | ||
NM_145239.2 | 154 | Intron | NP_660282.2 | ||
XM_011545715.2 | 154 | Intron | XP_011544017.1 | ||
XM_011545716.2 | 154 | Intron | XP_011544018.1 | ||
XM_017022887.1 | 154 | Intron | XP_016878376.1 | ||
XM_017022888.1 | 154 | Intron | XP_016878377.1 | ||
XM_017022889.1 | 154 | Intron | XP_016878378.1 |