Product Details

SNP ID
rs4788186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29829904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTACTAAAGCATAGGAAAGAGGC[G/A]GGGAACATAGACACAGCTCATGTAA
Phenotype
MIM: 605088 MIM: 612033
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MVP PubMed Links

Gene Details

Gene
MVP
Gene Name
major vault protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293204.1 Intron NP_001280133.1
NM_001293205.1 Intron NP_001280134.1
NM_005115.4 Intron NP_005106.2
NM_017458.3 Intron NP_059447.2
Gene
PAGR1
Gene Name
PAXIP1 associated glutamate rich protein 1
There are no transcripts associated with this gene.

View Full Product Details