Product Details

SNP ID
rs3815822
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:29861040 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTAGTGCCATCCCAGACACCCTTCC[G/A]TCTAGGCTCAGCCCACCCCACTGTA
Phenotype
MIM: 605893
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
CDIPT PubMed Links

Gene Details

Gene
CDIPT
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286585.1 Intron NP_001273514.1
NM_001286586.1 Intron NP_001273515.1
NM_006319.4 Intron NP_006310.1
XM_005255038.4 Intron XP_005255095.1
Gene
CDIPT-AS1
Gene Name
CDIPT antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details