Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001242338.2 | Intron | NP_001229267.2 | |||
NM_017552.3 | Intron | NP_060022.2 | |||
XM_005264372.3 | Intron | XP_005264429.2 | |||
XM_006712031.3 | Intron | XP_006712094.1 | |||
XM_011532918.2 | Intron | XP_011531220.1 | |||
XM_011532919.2 | Intron | XP_011531221.1 | |||
XM_011532920.2 | Intron | XP_011531222.1 | |||
XM_011532921.2 | Intron | XP_011531223.1 | |||
XM_011532924.2 | Intron | XP_011531226.1 | |||
XM_011532925.2 | Intron | XP_011531227.1 | |||
XM_011532928.1 | Intron | XP_011531230.1 | |||
XM_011532929.2 | Intron | XP_011531231.1 | |||
XM_011532930.2 | Intron | XP_011531232.1 | |||
XM_011532931.2 | Intron | XP_011531233.1 | |||
XM_011532932.1 | Intron | XP_011531234.1 | |||
XM_017004375.1 | Intron | XP_016859864.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_052920.1 | Intron | NP_443152.1 | |||
XM_006711929.3 | Intron | XP_006711992.1 | |||
XM_011532501.2 | Intron | XP_011530803.1 | |||
XM_017003264.1 | Intron | XP_016858753.1 |