Product Details
- SNP ID
-
rs12962202
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:35592865 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCATCATATGTATCATGTTACCCC[C/T]CTCTGTTTATCCTTGCCCCTTTTAG
- Phenotype
-
MIM: 602273
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GALNT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74442274] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GALNT1
- Gene Name
- polypeptide N-acetylgalactosaminyltransferase 1
- Gene
- MIR3975
- Gene Name
- microRNA 3975
There are no transcripts associated with this gene.
View Full Product Details