Product Details

SNP ID
rs981179
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:34977717 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GAGCCGGGTGGGTGCAAAAGGAAGC[A/G]GGTTGTTATTCTGCGCCCAAGCATC
Phenotype
MIM: 605789
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPRE2 PubMed Links

Gene Details

Gene
MAPRE2
Gene Name
microtubule associated protein RP/EB family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143826.2 Intron NP_001137298.1
NM_001143827.2 Intron NP_001137299.1
NM_001256420.1 Intron NP_001243349.1
NM_014268.3 Intron NP_055083.1

View Full Product Details