Product Details

SNP ID
rs12000845
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:72902146 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAGGAATTAACTCAGTCAATCAGT[C/T]TGGATTTTGGGATCTTGTTATCAGG
Phenotype
MIM: 100640
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH1A1 PubMed Links

Gene Details

Gene
ALDH1A1
Gene Name
aldehyde dehydrogenase 1 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000689.4 Intron NP_000680.2

View Full Product Details