Product Details
- SNP ID
-
rs7582124
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:159716326 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGTAAATAATGAAATAGTAACTGCG[C/T]GTTTGTATTTCTGCAAGACTCTTGA
- Phenotype
-
MIM: 605683
MIM: 613334
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BAZ2B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11454651] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BAZ2B
- Gene Name
- bromodomain adjacent to zinc finger domain 2B
There are no transcripts associated with this gene.
- Gene
- MARCH7
- Gene Name
- membrane associated ring-CH-type finger 7
View Full Product Details