Product Details
- SNP ID
-
rs6542397
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:117818654 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAAGCATTGAATCACAATGCTGGGC[A/G]TTTTAACCTTATGGGTTTTTTGTTT
- Phenotype
-
MIM: 606355
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DDX18
PubMed Links
Gene Details
- Gene
- DDX18
- Gene Name
- DEAD-box helicase 18
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006773.3 |
|
Intron |
|
|
NP_006764.3 |
View Full Product Details