Product Details

SNP ID
rs4724798
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:6344369 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAGACACCCCACTCCCCACCCCCA[T/C]GATGTGACATGGCTGCTTGTTGCAC
Phenotype
MIM: 616628
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
FAM220A PubMed Links

Gene Details

Gene
FAM220A
Gene Name
family with sequence similarity 220 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037163.1 Intron NP_001032240.1

View Full Product Details