Product Details

SNP ID
rs645900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65880664 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGAAATCGAAGCTCAGGGTGGG[A/G]AACAGGCATGGCCAAGGCCACTCAG
Phenotype
MIM: 605360 MIM: 602364 MIM: 604633 MIM: 608296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC85B PubMed Links

Gene Details

Gene
CCDC85B
Gene Name
coiled-coil domain containing 85B
There are no transcripts associated with this gene.

Gene
CTSW
Gene Name
cathepsin W
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001335.3 Intron NP_001326.2
Gene
EFEMP2
Gene Name
EGF containing fibulin like extracellular matrix protein 2
There are no transcripts associated with this gene.

Gene
FIBP
Gene Name
FGF1 intracellular binding protein
There are no transcripts associated with this gene.

View Full Product Details