Product Details

SNP ID
rs4793711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:51632443 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGAGAGAAAAAAGAAGGGAACA[A/T]GGGGGAAGTAGGAGGAATGAAGTGG
Phenotype
MIM: 604642
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CA10 PubMed Links

Gene Details

Gene
CA10
Gene Name
carbonic anhydrase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001082533.1 Intron NP_001076002.1
NM_001082534.1 Intron NP_001076003.1
NM_020178.4 Intron NP_064563.1
XM_017024878.1 Intron XP_016880367.1

View Full Product Details