Product Details

SNP ID
rs6663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109448798 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAACTGTTACAGTGACTTTGGGTAG[A/G]GCCCTAAAGACAGCACACGCTCCAG
Phenotype
MIM: 613421 MIM: 614636
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317395.1 4024 UTR 3 NP_001304324.1
NM_001317399.1 4024 UTR 3 NP_001304328.1
NM_031954.4 4024 UTR 3 NP_114160.1
Gene
MYO1H
Gene Name
myosin IH
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101421.3 4024 Intron NP_001094891.3
XM_011538223.2 4024 Intron XP_011536525.1
XM_017019207.1 4024 Intron XP_016874696.1

View Full Product Details