Product Details
- SNP ID
-
rs2303093
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:33209539 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTGCAGGGCCTCCAGCTGTCTGTA[C/T]CCCCGACCCCTGGGGGTCTGGGGAA
- Phenotype
-
MIM: 603159
MIM: 607959
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LRP3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78686000] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LRP3
- Gene Name
- LDL receptor related protein 3
- Gene
- SLC7A10
- Gene Name
- solute carrier family 7 member 10
View Full Product Details