Product Details

SNP ID
rs11671345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33199072 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGCCATCTTCTGCCTGAGAGAAG[G/T]CAGGTCACCTCCAGCTGAAGCTCAG
Phenotype
MIM: 603159 MIM: 607959
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LRP3 PubMed Links

Gene Details

Gene
LRP3
Gene Name
LDL receptor related protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002333.3 Intron NP_002324.2
XM_005258945.1 Intron XP_005259002.1
Gene
SLC7A10
Gene Name
solute carrier family 7 member 10
There are no transcripts associated with this gene.

View Full Product Details