Product Details

SNP ID
rs34140285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51382216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGGTGATTCATGCCAGAACTGAA[C/T]TGTAGTACACCAGCTAGTGTCAGAA
Phenotype
MIM: 154045 MIM: 606008
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C19orf84 PubMed Links

Gene Details

Gene
C19orf84
Gene Name
chromosome 19 open reading frame 84
There are no transcripts associated with this gene.

Gene
LIM2
Gene Name
lens intrinsic membrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161748.1 Intron NP_001155220.1
NM_030657.3 Intron NP_085915.2
Gene
NKG7
Gene Name
natural killer cell granule protein 7
There are no transcripts associated with this gene.

View Full Product Details