Product Details

SNP ID
rs2075588
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:38815785 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGGGCCAATCAGGATAAAGCATG[A/C]GAGCACCCTGCACCCTGGTTGGTCG
Phenotype
MIM: 600696 MIM: 602518
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ECH1 PubMed Links

Gene Details

Gene
ECH1
Gene Name
enoyl-CoA hydratase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001398.2 Intron NP_001389.2
XM_017026448.1 Intron XP_016881937.1
Gene
LGALS4
Gene Name
galectin 4
There are no transcripts associated with this gene.

View Full Product Details