Product Details

SNP ID
rs2135137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154791106 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACAGTTAAGTAGCATTTTCCCATG[A/G]TTCTGTAGAAACTTAATAGATAGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM46 PubMed Links

Gene Details

Gene
RBM46
Gene Name
RNA binding motif protein 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277171.1 Intron NP_001264100.1
NM_001277173.1 Intron NP_001264102.1
NM_144979.4 Intron NP_659416.1
XM_006714119.2 Intron XP_006714182.1
XM_006714120.1 Intron XP_006714183.1
XM_011531692.1 Intron XP_011529994.1
XM_011531693.1 Intron XP_011529995.1
XM_011531694.1 Intron XP_011529996.1
XM_011531695.2 Intron XP_011529997.1
XM_011531696.1 Intron XP_011529998.1
XM_011531697.2 Intron XP_011529999.1

View Full Product Details