Product Details

SNP ID
rs1876031
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:154741584 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCATCTTGAGCCACCTATACAGATT[C/G]ATGCCTCGTATCGCTCTCACCTCCT
Phenotype
MIM: 604863
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LRAT PubMed Links

Gene Details

Gene
LRAT
Gene Name
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301645.1 Intron NP_001288574.1
NM_004744.4 Intron NP_004735.2

View Full Product Details