Product Details
- SNP ID
-
rs1049636
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:154604818 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCTGAAACTTTGTGGGTCAATAGAA[A/G]TTAGCAGTTAATTTTCTACAAATCA
- Phenotype
-
MIM: 134850
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FGG
PubMed Links
Gene Details
- Gene
- FGG
- Gene Name
- fibrinogen gamma chain
View Full Product Details