Product Details
- SNP ID
-
rs4736991
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:41492136 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CAAATATGTTTCTTGAATTGCAAAT[G/T]GATCAAATATGCCTTTAAAAGATGG
- Phenotype
-
MIM: 609453
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA7
PubMed Links
Gene Details
- Gene
- GOLGA7
- Gene Name
- golgin A7
View Full Product Details