Product Details

SNP ID
rs933832
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41533235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGAGCTGCCAAAGAAACACTTT[A/C]TGTAAAGTTCAAACTTGCAAAATTA
Phenotype
MIM: 610611
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
GINS4 PubMed Links
Additional Information
For this assay, SNP(s) [rs75925561] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GINS4
Gene Name
GINS complex subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032336.2 Intron NP_115712.1
XM_005273659.3 Intron XP_005273716.1
Gene
LOC102723729
Gene Name
uncharacterized LOC102723729
There are no transcripts associated with this gene.

View Full Product Details