Product Details

SNP ID
rs35649072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:53228914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGGATGCCCATTGAGCTCTGAA[A/G]GCAATGTTTTTTCTTTTCCTTTTTT
Phenotype
MIM: 165196
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OPRK1 PubMed Links

Gene Details

Gene
OPRK1
Gene Name
opioid receptor kappa 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000912.4 1824 UTR 3 NP_000903.2
NM_001282904.1 1824 UTR 3 NP_001269833.1
NM_001318497.1 1824 UTR 3 NP_001305426.1

View Full Product Details