Product Details

SNP ID
rs709564
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:107377700 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AATGTTTACCACAAATGTAAGATTC[A/G]GCACCAAGATTCAACTGGATATCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC54 PubMed Links

Gene Details

Gene
CCDC54
Gene Name
coiled-coil domain containing 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032600.2 360 Missense Mutation CAG,CGG Q38R NP_115989.1
Gene
LOC101929579
Gene Name
uncharacterized LOC101929579
There are no transcripts associated with this gene.

View Full Product Details