Product Details

SNP ID
rs3889590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73458140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCCCAATTCTGAAGAAAGGGTGG[T/C]AGGGAAGAGGGGACAATTCACTAAG
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
REC114 PubMed Links

Gene Details

Gene
REC114
Gene Name
REC114 meiotic recombination protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042367.1 Intron NP_001035826.1
XM_017022083.1 Intron XP_016877572.1

View Full Product Details