Product Details

SNP ID
rs279940
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:103046280 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAAACTAGAAAACAATACACAGA[C/T]AGTTAAGTAAATTTTATAATAATAA
Phenotype
MIM: 601295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC10A2 PubMed Links

Gene Details

Gene
SLC10A2
Gene Name
solute carrier family 10 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000452.2 Intron NP_000443.1

View Full Product Details